Gene Transcripts
- Gene Symbol
- 2410114N07Rik
- Entrez Gene ID
- 73696
- Gene Name
- RIKEN cDNA 2410114N07 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 34909789 - 34911481 on Build GRCm38
- UniGene ID
- Mm.1361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040652.1 |
|
3 |
797 |
|
GenBank mRNA |
AK010767.1 |
- |
3 |
798 |
|
View Full Product Details