Gene Transcripts
- Gene Symbol
- LINC01252
- Entrez Gene ID
- 338817
- Gene Name
- long intergenic non-protein coding RNA 1252
- Gene Aliases
- Chromosome Location
- Chr. 12: 11548030 - 11564403 on Build GRCh38
- UniGene ID
- Hs.733066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033890.1 |
|
7 |
2920 |
|
GenBank mRNA |
BC041827.1 |
- |
7 |
2923 |
|
View Full Product Details