Gene Transcripts
- Gene Symbol
- Olfr883
- Entrez Gene ID
- 258414
- Gene Name
- olfactory receptor 883
- Gene Aliases
- MOR162-6
- Chromosome Location
- Chr. 9: 38025808 - 38026737 on Build GRCm38
- UniGene ID
- Mm.377487
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146419.2 |
NP_666531.1
|
1 |
389 |
|
View Full Product Details