Gene Transcripts

Gene Symbol
Tgm4
Entrez Gene ID
64679
Gene Name
transglutaminase 4
Gene Aliases
Dp1
Chromosome Location
Chr. 8: 132117273 - 132157485 on Build Rnor_6.0
UniGene ID
Rn.9964
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022713.2 NP_073204.2 10 1331
XM_006244202.2 XP_006244264.1 10 1511
GenBank mRNA BC066665.1 - 10 1320
M32725.1 - 6 1032

View Full Product Details