Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297438.1 | NP_001284367.1 | 3 | 602 | |
NM_014231.4 | NP_055046.1 | 3 | 602 | ||
NM_016830.3 | NP_058439.1 | 3 | 602 | ||
NM_199245.2 | NP_954740.1 | 3 | 602 | ||
NR_123717.1 | 2 | 475 | |||
GenBank mRNA | AF060538.1 | - | 3 | 364 | |
AK289498.1 | - | 3 | 373 | ||
AK296294.1 | - | 3 | 380 | ||
AK303137.1 | - | 3 | 380 | ||
AK309184.1 | - | 2 | 186 | ||
BC023286.1 | - | 3 | 368 | 4943992 | |
BE797045.1 | - | 2 | 190 | ||
BI838752.1 | - | 3 | 397 | ||
BQ717992.1 | - | 3 | 361 | ||
CR542214.1 | - | 2 | 234 | ||
CR542231.1 | - | 2 | 234 | ||
CR749485.1 | - | 3 | 603 | ||
CR989458.1 | - | 3 | 356 | ||
Z48924.1 | - | 2 | 234 |