Gene Transcripts
- Gene Symbol
- Mamdc4
- Entrez Gene ID
- 252882
- Gene Name
- MAM domain containing 4
- Gene Aliases
- Aegp
- Chromosome Location
- Chr. 3: 2786523 - 2795109 on Build Rnor_6.0
- UniGene ID
- Rn.10059
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145768.1 |
NP_665711.1
|
17 |
2118 |
|
GenBank mRNA |
L37380.1 |
- |
17 |
2118 |
|
View Full Product Details