Gene Transcripts

Gene Symbol
Map3k7cl
Entrez Gene ID
304131
Gene Name
MAP3K7 C-terminal like
Gene Aliases
Chromosome Location
Chr. 11: 27208149 - 27251802 on Build Rnor_6.0
UniGene ID
Rn.32787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013979.1 NP_001014001.1 4 484
XM_006248016.2 XP_006248078.1 5 1196
GenBank mRNA BC076389.1 - 4 484

View Full Product Details