Gene Transcripts

Gene Symbol
Tfcp2
Entrez Gene ID
315309
Gene Name
transcription factor CP2
Gene Aliases
Tcfcp2
Chromosome Location
Chr. 7: 142138843 - 142181034 on Build Rnor_6.0
UniGene ID
Rn.202290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134714.1 NP_001128186.1 6 981
XM_006242341.2 XP_006242403.1 7 822
GenBank mRNA BC168176.1 - 6 981

View Full Product Details