Gene Transcripts
- Gene Symbol
- Traf7
- Entrez Gene ID
- 360491
- Gene Name
- TNF receptor-associated factor 7, E3 ubiquitin protein ligase
- Gene Aliases
- RGD1559653
- Chromosome Location
- Chr. 10: 13874456 - 13893091 on Build Rnor_6.0
- UniGene ID
- Rn.163128
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127548.1 |
NP_001121020.1
|
15, 16 |
1481 |
|
XM_006245982.2 |
XP_006246044.1
|
15, 16 |
1575 |
|
XM_006245983.2 |
XP_006246045.1
|
15, 16 |
1570 |
|
XM_006245984.2 |
XP_006246046.1
|
14, 15 |
1416 |
|
XM_006245985.2 |
XP_006246047.1
|
14, 15 |
1393 |
|
XM_006245986.2 |
XP_006246048.1
|
14, 15 |
1413 |
|
XM_006245987.2 |
XP_006246049.1
|
14, 15 |
1390 |
|
XM_006245988.2 |
XP_006246050.1
|
15, 16 |
1612 |
|
XM_006245989.2 |
XP_006246051.1
|
15, 16 |
1609 |
|
XM_006245990.2 |
XP_006246052.1
|
14, 15 |
1362 |
|
XM_006245991.2 |
XP_006246053.1
|
14, 15 |
1366 |
|
XM_006245992.2 |
XP_006246054.1
|
14, 15 |
1359 |
|
XM_006245993.2 |
XP_006246055.1
|
13, 14 |
1352 |
|
XM_008767584.1 |
XP_008765806.1
|
15, 16 |
1628 |
|
XM_008767585.1 |
XP_008765807.1
|
15, 16 |
1625 |
|
XM_008767586.1 |
XP_008765808.1
|
15, 16 |
1623 |
|
GenBank mRNA |
BC161916.1 |
- |
15, 16 |
1481 |
|
View Full Product Details