Gene Transcripts

Gene Symbol
LOC691030
Entrez Gene ID
691030
Gene Name
similar to RIKEN cDNA 1700001F22
Gene Aliases
Chromosome Location
Chr. X: 130794079 - 130794811 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773536.1 XP_008771758.1 1 642

View Full Product Details