Gene Transcripts
- Gene Symbol
- FAM153A
- Entrez Gene ID
- 285596
- Gene Name
- family with sequence similarity 153 member A
- Gene Aliases
- NY-REN-7
- Chromosome Location
- Chr. 5: 177694247 - 177783518 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
- Gene Symbol
- FAM153B
- Entrez Gene ID
- 202134
- Gene Name
- family with sequence similarity 153 member B
- Gene Aliases
- Chromosome Location
- Chr. 5: 176063709 - 176114798 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL042735.1 |
- |
3 |
186 |
|
DB291737.1 |
- |
4 |
455 |
|
- Gene Symbol
- FAM153C
- Entrez Gene ID
- 653316
- Gene Name
- family with sequence similarity 153, member C
- Gene Aliases
- NY-REN-7-like
- Chromosome Location
- Chr. 5: 178006966 - 178049087 on Build GRCh38
- UniGene ID
- Hs.652193
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038353.1 |
|
12 |
708 |
|
GenBank mRNA |
AK294762.1 |
- |
12 |
702 |
|
DA083707.1 |
- |
5 |
301 |
|
View Full Product Details