Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
234396
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
8430438L13Rik, Ankrd41, C430003I23, Lem3
Chromosome Location
Chr. 8: 71406005 - 71410543 on Build GRCm38
UniGene ID
Mm.216549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172756.2 NP_766344.2 6 1139
GenBank mRNA AK049395.1 - 4 1446
AK078830.1 - 5 1369
AK080768.1 - 4 1452
AK083552.1 - 4 1438
AK087444.1 - 4 1457
BC030436.1 - 6 1078
BC094316.1 - 6 1096
BC145659.1 - 2 362
EU184014.1 - 6 1115

View Full Product Details