Gene Transcripts

Gene Symbol
Zfp661
Entrez Gene ID
72180
Gene Name
zinc finger protein 661
Gene Aliases
2810405K07Rik, A1-5, Znf2
Chromosome Location
Chr. 2: 127574617 - 127587135 on Build GRCm38
UniGene ID
Mm.46715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111029.1 NP_001104499.1 5 573
NM_028141.3 NP_082417.3 6 790
GenBank mRNA AK033122.1 - 5 575
BC052876.1 - 6 794 30047253

View Full Product Details