Gene Transcripts

Gene Symbol
TTC23L
Entrez Gene ID
153657
Gene Name
tetratricopeptide repeat domain 23 like
Gene Aliases
MC25-1
Chromosome Location
Chr. 5: 34839164 - 34900511 on Build GRCh38
UniGene ID
Hs.435742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317949.1 NP_001304878.1 8, 9 994
NM_144725.3 NP_653326.3 8, 9 1059
XM_011513984.2 XP_011512286.1 8, 9 996
XM_011513985.2 XP_011512287.1 8, 9 996
XM_017009119.1 XP_016864608.1 8, 9 1240
XM_017009120.1 XP_016864609.1 8, 9 1059
XM_017009121.1 XP_016864610.1 8, 9 1104
XM_017009122.1 XP_016864611.1 8, 9 1175
XM_017009124.1 XP_016864613.1 6, 7 936
XM_017009125.1 XP_016864614.1 6, 7 737
XM_017009126.1 XP_016864615.1 6, 7 870
GenBank mRNA AK058168.1 - 8, 9 1059
BC057775.1 - 8, 9 1031 5295348
BX088767.1 - 3, 4 336
JN799997.1 - 8, 9 994

View Full Product Details