Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr. 10: 22928024 - 23038527 on Build GRCh38
UniGene ID
Hs.659807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282745.1 NP_001269674.1 14 1904
NM_001282746.1 NP_001269675.1 14 1904
NM_001282747.1 NP_001269676.1 11 1415
NM_173081.4 NP_775104.2 14 1904
XM_005252380.2 XP_005252437.1 13 1692
XM_005252381.2 XP_005252438.1 13 1725
XM_011519350.2 XP_011517652.1 14 1887
XM_011519351.2 XP_011517653.1 5 734
XM_017015831.1 XP_016871320.1 13 1692
XM_017015832.1 XP_016871321.1 14 1887
XM_017015833.1 XP_016871322.1 13 1692
GenBank mRNA AK057389.1 - 14 1847
AK098711.1 - 4 444
AK302104.1 - 11 1362
AK302985.1 - 14 1891
AY436643.1 - 14 1865
BC039312.1 - 14 1897 5296302

View Full Product Details