Gene Transcripts
- Gene Symbol
- C1orf111
- Entrez Gene ID
- 284680
- Gene Name
- chromosome 1 open reading frame 111
- Gene Aliases
- HSD20
- Chromosome Location
- Chr. 1: 162373725 - 162376962 on Build GRCh38
- UniGene ID
- Hs.97784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC032957.1 |
- |
3 |
711 |
|
View Full Product Details