Gene Transcripts

Gene Symbol
LINC01630
Entrez Gene ID
100287225
Gene Name
long intergenic non-protein coding RNA 1630
Gene Aliases
Chromosome Location
Chr. 18: 51392042 - 51562469 on Build GRCh38
UniGene ID
Hs.448920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040074.1 1 179
NR_040075.1 1 179
GenBank mRNA AK094936.1 - 1 179
AK294844.1 - 1 179

View Full Product Details