Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010240.2 | NP_034370.2 | 4 | 872 | |
GenBank mRNA | AK002242.1 | - | 4 | 826 | |
AK002253.1 | - | 4 | 825 | ||
AK011009.1 | - | 4 | 825 | ||
AK011029.1 | - | 4 | 824 | ||
AK011244.1 | - | 4 | 826 | ||
AK012690.1 | - | 3 | 411 | ||
AK088647.1 | - | 4 | 830 | ||
AK145265.1 | - | 4 | 825 | ||
AK145658.1 | - | 4 | 825 | ||
AK150480.1 | - | 4 | 825 | ||
AK150481.1 | - | 4 | 821 | ||
AK150960.1 | - | 4 | 847 | ||
AK152030.1 | - | 4 | 825 | ||
AK152385.1 | - | 4 | 821 | ||
AK152393.1 | - | 4 | 823 | ||
AK152564.1 | - | 4 | 825 | ||
AK152612.1 | - | 4 | 825 | ||
AK159574.1 | - | 4 | 826 | ||
AK167410.1 | - | 4 | 823 | ||
AK168310.1 | - | 4 | 825 | ||
AK168735.1 | - | 4 | 825 | ||
AK168862.1 | - | 4 | 826 | ||
AK168866.1 | - | 4 | 825 | ||
AK169100.1 | - | 4 | 825 | ||
AK169159.1 | - | 4 | 826 | ||
BC019840.1 | - | 4 | 804 | 5123770 | |
BC020403.1 | - | 4 | 813 | ||
BC081462.1 | - | 4 | 791 | 6823712 | |
BC083350.1 | - | 4 | 802 | 6824407 | |
BC085309.1 | - | 4 | 804 | 6489467 | |
BC092259.1 | - | 4 | 793 | 5125514 | |
BC106145.1 | - | 4 | 791 | 5068947 | |
BC106146.1 | - | 4 | 804 | 6476394 | |
J04716.1 | - | 4 | 791 |