Gene Transcripts
- Gene Symbol
- Ifna1
- Entrez Gene ID
- 15962
- Gene Name
- interferon alpha 1
- Gene Aliases
- Ifa1
- Chromosome Location
- Chr. 4: 88850087 - 88850656 on Build GRCm38
- UniGene ID
- Mm.57127
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010502.2 |
NP_034632.2
|
1 |
87 |
|
- Gene Symbol
- Ifna5
- Entrez Gene ID
- 15968
- Gene Name
- interferon alpha 5
- Gene Aliases
- Ifa5
- Chromosome Location
- Chr. 4: 88835525 - 88836094 on Build GRCm38
- UniGene ID
- Mm.377089
- Species
- Mus musculus
- Gene Symbol
- Ifna6
- Entrez Gene ID
- 15969
- Gene Name
- interferon alpha 6
- Gene Aliases
- IFNa8/6, Ifa6, Ifa8, Ifna8
- Chromosome Location
- Chr. 4: 88827416 - 88827985 on Build GRCm38
- UniGene ID
- Mm.377090
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC117774.1 |
- |
|
87 |
|
- Gene Symbol
- Ifna7
- Entrez Gene ID
- 15970
- Gene Name
- interferon alpha 7
- Gene Aliases
- Ifa7
- Chromosome Location
- Chr. 4: 88816225 - 88816800 on Build GRCm38
- UniGene ID
- Mm.377091
- Species
- Mus musculus
View Full Product Details