Gene Transcripts

Gene Symbol
Ifna4
Entrez Gene ID
15967
Gene Name
interferon alpha 4
Gene Aliases
Ifa4
Chromosome Location
Chr. 4: 88841816 - 88842459 on Build GRCm38
UniGene ID
Mm.377088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010504.2 NP_034634.1 1 288
GenBank mRNA BC116217.2 - 1 243
BC119349.1 - 1 288 8734098
BC119351.1 - 1 288 8734100

View Full Product Details