Gene Transcripts

Gene Symbol
1600014K23Rik
Entrez Gene ID
71996
Gene Name
RIKEN cDNA 1600014K23 gene
Gene Aliases
AI324104
Chromosome Location
Chr. X: 85249677 - 85270291 on Build GRCm38
UniGene ID
Mm.20201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028046.1 NP_082322.1 2 761
GenBank mRNA AK005459.1 - 1 719
BC049727.1 - 2 752 6770312

View Full Product Details