Gene Transcripts
- Gene Symbol
- Zfp661
- Entrez Gene ID
- 72180
- Gene Name
- zinc finger protein 661
- Gene Aliases
- 2810405K07Rik, A1-5, Znf2
- Chromosome Location
- Chr. 2: 127574617 - 127587135 on Build GRCm38
- UniGene ID
- Mm.46715
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052876.1 |
- |
6 |
597 |
|
View Full Product Details