Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr. 1: 180165238 - 180196020 on Build GRCm38
UniGene ID
Mm.38330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163290.1 NP_001156762.1 7 960
NM_023341.3 NP_075830.2 7 1058
GenBank mRNA AJ278735.1 - 7 938
AK004791.1 - 7 1000
AK014605.2 - 7 1000
AK184581.1 - 4 332
BC030937.1 - 7 1006 4922892
U31629.1 - 5 599

View Full Product Details