Gene Transcripts

Gene Symbol
Thap7
Entrez Gene ID
69009
Gene Name
THAP domain containing 7
Gene Aliases
1810004B07Rik
Chromosome Location
Chr. 16: 17527558 - 17531512 on Build GRCm38
UniGene ID
Mm.10174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026909.2 NP_081185.1 5 430
GenBank mRNA AK007329.1 - 4 382
AK154641.1 - 5 516
BC018221.1 - 5 430 4215606

View Full Product Details