Gene Transcripts
- Gene Symbol
- Chn1
- Entrez Gene ID
- 108699
- Gene Name
- chimerin 1
- Gene Aliases
- 0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2
- Chromosome Location
- Chr. 2: 73610660 - 73775367 on Build GRCm38
- UniGene ID
- Mm.257073
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113246.2 |
NP_001106717.2
|
9, 10 |
1199 |
|
NM_029716.3 |
NP_083992.1
|
2, 3 |
400 |
|
NM_175752.3 |
NP_786928.2
|
3, 4 |
1011 |
|
GenBank mRNA |
AB264774.1 |
- |
1, 2 |
801 |
|
AF332069.1 |
- |
3, 4 |
519 |
|
AF332070.1 |
- |
3, 4 |
519 |
|
AK007182.1 |
- |
2, 3 |
400 |
|
AK032630.1 |
- |
3, 4 |
969 |
|
AK049943.1 |
- |
3, 4 |
955 |
|
AK075606.1 |
- |
3, 4 |
973 |
|
AK147686.1 |
- |
3, 4 |
955 |
|
BC010825.1 |
- |
3, 4 |
986 |
4219273 |
BC024796.1 |
- |
3, 4 |
962 |
5360438 |
BC025023.1 |
- |
3, 4 |
942 |
5364554 |
BC054770.1 |
- |
9, 10 |
951 |
6415204 |
View Full Product Details