Gene Transcripts

Gene Symbol
Ins2
Entrez Gene ID
16334
Gene Name
insulin II
Gene Aliases
AA986540, Ins-2, InsII, Mody, Mody4, proinsulin
Chromosome Location
Chr. 7: 142678656 - 142679708 on Build GRCm38
UniGene ID
Mm.4946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185083.1 NP_001172012.1 2 150
NM_001185084.1 NP_001172013.1 1 264
NM_008387.4 NP_032413.1 2 162
GenBank mRNA AK007612.1 - 2 163
AY899305.1 - 1 229
BC066208.1 - 2 127
BC099934.1 - 2 135 6432976
BC132650.1 - 2 135 40130927
BC132652.1 - 2 135 40130929
BC145554.1 - 2 123 9054118
GQ915612.1 - 2 150

View Full Product Details