Gene Transcripts

Gene Symbol
Alyref2
Entrez Gene ID
56009
Gene Name
Aly/REF export factor 2
Gene Aliases
C130042O11Rik, REF2, Refbp2
Chromosome Location
Chr. 1: 171503478 - 171504750 on Build GRCm38
UniGene ID
Mm.389208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019484.4 NP_062357.3 1 327
GenBank mRNA AJ252141.1 - 1 148
AK048243.1 - 1 328
BC099405.1 - 1 234 30846460

View Full Product Details