Gene Transcripts

Gene Symbol
Ypel3
Entrez Gene ID
66090
Gene Name
yippee-like 3 (Drosophila)
Gene Aliases
0610043B10Rik, 1190001G19Rik, Suap
Chromosome Location
Chr. 7: 126776975 - 126780514 on Build GRCm38
UniGene ID
Mm.27114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025347.2 NP_079623.1 5 480
NM_026875.2 NP_081151.2 5 580
GenBank mRNA AB098742.1 - 4 285
AF523353.1 - 5 470
AK002925.1 - 5 465
AK003371.1 - 5 481
AK004431.1 - 5 553
AK210119.1 - 4 260
BC009171.1 - 5 454

View Full Product Details