Gene Transcripts

Gene Symbol
Slc19a3
Entrez Gene ID
80721
Gene Name
solute carrier family 19, member 3
Gene Aliases
A230084E24Rik, AI788884, ThTr2
Chromosome Location
Chr. 1: 83012523 - 83038859 on Build GRCm38
UniGene ID
Mm.261542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030556.2 NP_085033.2 4 582
GenBank mRNA AF271634.1 - 4 582
BC109154.1 - 4 565 40047017
BC109155.1 - 3 435 40047019

View Full Product Details