Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031248.3 | NP_112538.1 | 2 | 275 | |
GenBank mRNA | AJ277386.1 | - | 2 | 245 | |
AK008126.1 | - | 2 | 292 | ||
AK008185.1 | - | 2 | 295 | ||
AK008395.1 | - | 2 | 292 | ||
AK019006.1 | - | 2 | 270 | ||
BC031816.1 | - | 2 | 275 | 4189054 |