Gene Transcripts
- Gene Symbol
- Cmtm7
- Entrez Gene ID
- 102545
- Gene Name
- CKLF-like MARVEL transmembrane domain containing 7
- Gene Aliases
- AI481279, Cklfsf7, LNV
- Chromosome Location
- Chr. 9: 114756836 - 114781993 on Build GRCm38
- UniGene ID
- Mm.35600
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252479.1 |
NP_001239408.1
|
1, 2 |
331 |
|
NM_133978.2 |
NP_598739.1
|
1, 2 |
331 |
|
GenBank mRNA |
AF188504.1 |
- |
1, 2 |
167 |
|
AK040123.1 |
- |
1, 2 |
195 |
|
AK151785.1 |
- |
1, 2 |
134 |
|
AK167886.1 |
- |
1, 2 |
177 |
|
AK168024.1 |
- |
1, 2 |
184 |
|
AK168280.1 |
- |
1, 2 |
168 |
|
AY241869.1 |
- |
1, 2 |
188 |
|
BC038633.1 |
- |
1, 2 |
170 |
1066813 |
View Full Product Details