Gene Transcripts

Gene Symbol
Olfr25
Entrez Gene ID
18323
Gene Name
olfactory receptor 25
Gene Aliases
MOR170-4, MTPCR18
Chromosome Location
Chr. 9: 38329492 - 38330559 on Build GRCm38
UniGene ID
Mm.222835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146870.2 NP_667081.2 1 169
GenBank mRNA BC125306.1 - 1 169
BC125308.1 - 1 169 40129823

View Full Product Details