Gene Transcripts

Gene Symbol
STK17B
Entrez Gene ID
9262
Gene Name
serine/threonine kinase 17b
Gene Aliases
DRAK2
Chromosome Location
Chr. 2: 196133583 - 196176503 on Build GRCh38
UniGene ID
Hs.88297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB011421.1 - 3 482
AK312865.1 - 3 543
BC016040.1 - 3 484 4575012

View Full Product Details