Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001713.2 | NP_001704.2 | 8 | 1609 | |
GenBank mRNA | AK303780.1 | - | 5 | 1121 | |
BC012616.1 | - | 8 | 1585 | 4045543 | |
EU794592.1 | - | 8 | 1609 | ||
U50929.1 | - | 8 | 1530 |