Gene Transcripts
- Gene Symbol
- PPM1D
- Entrez Gene ID
- 8493
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1D
- Gene Aliases
- PP2C-DELTA, WIP1
- Chromosome Location
- Chr. 17: 60600183 - 60666280 on Build GRCh38
- UniGene ID
- Hs.286073
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003620.3 |
NP_003611.1
|
2, 3 |
928 |
|
GenBank mRNA |
AK299773.1 |
- |
3, 4 |
637 |
|
AK312902.1 |
- |
2, 3 |
918 |
|
BC016480.1 |
- |
2, 3 |
844 |
3923593 |
BC033893.2 |
- |
2, 3 |
802 |
5265521 |
BC042418.1 |
- |
2, 3 |
842 |
5167004 |
BC060877.1 |
- |
2, 3 |
928 |
5260081 |
BT009780.1 |
- |
2, 3 |
696 |
|
KU178420.1 |
- |
2, 3 |
696 |
|
U78305.1 |
- |
2, 3 |
899 |
|
View Full Product Details