Gene Transcripts

Gene Symbol
ASIC5
Entrez Gene ID
51802
Gene Name
acid sensing ion channel subunit family member 5
Gene Aliases
ACCN5, HINAC, INAC
Chromosome Location
Chr. 4: 155829699 - 155867303 on Build GRCh38
UniGene ID
Hs.381349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017419.2 NP_059115.1 5 894
XM_017008291.1 XP_016863780.1 4 1798
GenBank mRNA AJ252011.1 - 5 894

View Full Product Details