Gene Transcripts
- Gene Symbol
- USP6
- Entrez Gene ID
- 9098
- Gene Name
- ubiquitin specific peptidase 6
- Gene Aliases
- HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
- Chromosome Location
- Chr. 17: 5116438 - 5175032 on Build GRCh38
- UniGene ID
- Hs.448851
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304284.1 |
NP_001291213.1
|
31, 32 |
5061 |
|
NM_004505.3 |
NP_004496.2
|
23, 24 |
4530 |
|
XM_011524050.1 |
XP_011522352.1
|
27, 28 |
5279 |
|
XM_011524051.2 |
XP_011522353.1
|
29, 30 |
4374 |
|
XM_011524052.2 |
XP_011522354.1
|
28, 29 |
4488 |
|
XM_011524053.2 |
XP_011522355.1
|
29, 30 |
4401 |
|
XM_011524054.2 |
XP_011522356.1
|
29, 30 |
4368 |
|
XM_011524055.2 |
XP_011522357.1
|
27, 28 |
4295 |
|
XM_011524056.2 |
XP_011522358.1
|
24, 25 |
4248 |
|
XM_011524057.2 |
XP_011522359.1
|
27, 28 |
4411 |
|
XM_011524058.2 |
XP_011522360.1
|
28, 29 |
4477 |
|
XM_017025290.1 |
XP_016880779.1
|
26, 27 |
5117 |
|
GenBank mRNA |
AY143550.1 |
- |
23, 24 |
4530 |
|
BX647719.1 |
- |
31, 32 |
5063 |
|
X63546.1 |
- |
22, 23 |
4415 |
|
X63547.1 |
- |
24, 25 |
4738 |
|
View Full Product Details