Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001860.2 | NP_001851.1 | 3 | 249 | |
XM_011518216.1 | XP_011516518.1 | 4 | 715 | ||
XM_017014267.1 | XP_016869756.1 | 4 | 4753 | ||
GenBank mRNA | AK057903.1 | - | 3 | 200 | |
AK131071.1 | - | 2 | 1873 | ||
AK131546.1 | - | 3 | 1739 | ||
AK295544.1 | - | 3 | 195 | ||
BC026252.1 | - | 3 | 195 | 4731026 | |
BP341248.1 | - | 3 | 249 | ||
CR407615.1 | - | 2 | 132 | ||
U83461.1 | - | 3 | 195 |