Gene Transcripts

Gene Symbol
GTF3A
Entrez Gene ID
2971
Gene Name
general transcription factor IIIA
Gene Aliases
AP2, TFIIIA
Chromosome Location
Chr. 13: 27424544 - 27435709 on Build GRCh38
UniGene ID
Hs.445977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002097.2 NP_002088.2 7 956
GenBank mRNA AK057993.1 - 7 1492
BC143711.1 - 6 948
D32257.1 - 7 955
U14134.1 - 6 681
U20272.1 - 7 756

View Full Product Details