Gene Transcripts

Gene Symbol
ROBO1
Entrez Gene ID
6091
Gene Name
roundabout guidance receptor 1
Gene Aliases
DUTT1, SAX3
Chromosome Location
Chr. 3: 78597238 - 79767998 on Build GRCh38
UniGene ID
Hs.744218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145845.1 NP_001139317.1 29 5675
NM_002941.3 NP_002932.1 31 5185
NM_133631.3 NP_598334.2 29 5840
XM_006713277.2 XP_006713340.1 30 5836
XM_011533976.1 XP_011532278.1 32 5283
XM_011533977.2 XP_011532279.1 32 5136
XM_011533978.1 XP_011532280.1 31 5256
XM_011533979.1 XP_011532281.1 32 5118
XM_011533980.1 XP_011532282.1 31 5091
XM_017006982.1 XP_016862471.1 31 5195
XM_017006983.1 XP_016862472.1 29 5827
XM_017006984.1 XP_016862473.1 31 5109
XM_017006985.1 XP_016862474.1 30 4936
GenBank mRNA AK025535.1 - 5 940
BC001969.1 - 7 1508
BC112336.1 - 29 4896 40037251
BC115020.1 - 29 4941 7961997
BC115022.1 - 29 4776 7962000
BC157861.1 - 29 5137 9057080
BC171855.1 - 29 5137 9054509
BX538319.1 - 23 3896

View Full Product Details