Gene Transcripts

Gene Symbol
CHGB
Entrez Gene ID
1114
Gene Name
chromogranin B
Gene Aliases
SCG1
Chromosome Location
Chr. 20: 5911328 - 5925361 on Build GRCh38
UniGene ID
Hs.516874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001819.2 NP_001810.2 4 926
GenBank mRNA AB209712.1 - 4 723
AK225125.1 - 4 744
AK289386.1 - 4 746
AK293536.1 - 4 480
AK312237.1 - 4 746
BC000375.2 - 4 724 2820258
BP422638.1 - 1 124
CR456726.1 - 4 620
Y00064.1 - 4 732

View Full Product Details