Gene Transcripts

Gene Symbol
SLC16A6
Entrez Gene ID
9120
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT6, MCT7
Chromosome Location
Chr. 17: 68267026 - 68291264 on Build GRCh38
UniGene ID
Hs.42645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174166.1 NP_001167637.1 6 1379
NM_004694.4 NP_004685.2 5 1252
XM_005257789.3 XP_005257846.2 5 1417
XM_011525461.2 XP_011523763.1 5 1096
XM_017025291.1 XP_016880780.1 6 1216
XM_017025292.1 XP_016880781.1 5 1279
GenBank mRNA AK290417.1 - 5 1128
AK313121.1 - 5 1118
AX750581.1 - 6 1231
AX750582.1 - 4 1066
BC064832.1 - 5 1254 6498761
BC127749.1 - 1 116
BC127750.1 - 1 114
BC141962.1 - 1 115
BC150547.1 - 1 116
BC150548.1 - 1 116
U79745.1 - 6 1231

View Full Product Details