Gene Transcripts

Gene Symbol
GNPDA1
Entrez Gene ID
10007
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
GNP1, GNPDA, GNPI, GPI, HLN
Chromosome Location
Chr. 5: 142000669 - 142013055 on Build GRCh38
UniGene ID
Hs.633853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005471.4 NP_005462.1 7 1814
XM_005268348.1 XP_005268405.1 7 2071
XM_006714747.2 XP_006714810.1 8 1838
XM_011537537.1 XP_011535839.1 8 2133
GenBank mRNA AF029914.1 - 6 1750
AJ002231.1 - 6 2496
AK022779.1 - 6 1674
BC001717.1 - 1 504
BC022322.1 - 7 1776 4243999
D31766.1 - 8 2138

View Full Product Details