Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005996.3 | NP_005987.3 | 6 | 2151 | |
NM_016569.3 | NP_057653.3 | 7 | 2211 | ||
GenBank mRNA | AF002228.1 | - | 6 | 1187 | |
AF170708.2 | - | 6 | 1447 | ||
AF216750.1 | - | 7 | 1363 | ||
BC025258.1 | - | 7 | 1637 | 5458526 |