Gene Transcripts
- Gene Symbol
- TAF5
- Entrez Gene ID
- 6877
- Gene Name
- TATA-box binding protein associated factor 5
- Gene Aliases
- TAF(II)100, TAF2D, TAFII-100, TAFII100
- Chromosome Location
- Chr. 10: 103367953 - 103389065 on Build GRCh38
- UniGene ID
- Hs.96103
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006951.4 |
NP_008882.2
|
9, 10 |
2052 |
|
NM_139052.2 |
NP_620640.1
|
8, 9 |
1887 |
|
GenBank mRNA |
AK223615.1 |
- |
9, 10 |
2024 |
|
AK291229.1 |
- |
9, 10 |
2052 |
|
BC052268.1 |
- |
9, 10 |
2030 |
6575596 |
BC136340.1 |
- |
8, 9 |
1868 |
9020327 |
BC136348.1 |
- |
9, 10 |
2033 |
9020335 |
U75309.1 |
- |
9, 10 |
2018 |
|
U80191.1 |
- |
9, 10 |
2029 |
|
X95525.1 |
- |
9, 10 |
2038 |
|
View Full Product Details