Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282692.1 | NP_001269621.1 | 2 | 321 | |
NM_001282693.1 | NP_001269622.1 | 3 | 315 | ||
NM_002021.2 | NP_002012.1 | 3 | 306 | ||
XM_005245038.3 | XP_005245095.1 | 3 | 315 | ||
XM_006711241.3 | XP_006711304.1 | 3 | 315 | ||
XM_006711242.3 | XP_006711305.1 | 3 | 315 | ||
GenBank mRNA | AK097039.1 | - | 3 | 275 | |
AK290113.1 | - | 3 | 292 | ||
AK315100.1 | - | 3 | 265 | ||
BC047129.1 | - | 2 | 287 | 5186727 | |
DA537896.1 | - | 3 | 316 | ||
DA750161.1 | - | 2 | 321 | ||
M64082.1 | - | 3 | 254 |