Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278940.1 | NP_001265869.1 | 8 | 1197 | |
NM_015859.3 | NP_056943.1 | 7 | 1130 | ||
NM_201595.2 | NP_963889.1 | 7 | 750 | ||
GenBank mRNA | BC107155.2 | - | 7 | 865 | |
BC107156.1 | - | 7 | 867 | 40032650 | |
D14886.1 | - | 7 | 750 | ||
D14887.1 | - | 7 | 1100 | ||
X75383.1 | - | 7 | 837 | ||
X77225.1 | - | 7 | 696 |