Gene Transcripts

Gene Symbol
FLVCR2
Entrez Gene ID
55640
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Gene Aliases
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH
Chromosome Location
Chr. 14: 75578597 - 75648169 on Build GRCh38
UniGene ID
Hs.509966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195283.1 NP_001182212.1 8 873
NM_017791.2 NP_060261.2 8 1755
GenBank mRNA AF456126.1 - 8 1379
AK000378.1 - 8 1703
AK027804.1 - 7 643
AK297002.1 - 8 873
AY260572.1 - 8 1735
AY260577.1 - 8 1735
BC019087.2 - 8 1718 4866427

View Full Product Details