Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252148.1 | NP_001239077.1 | 6 | 3369 | |
NM_001252151.1 | NP_001239080.1 | 7 | 2865 | ||
NM_001252152.1 | NP_001239081.1 | 8 | 2853 | ||
NM_018375.4 | NP_060845.2 | 7 | 3438 | ||
XM_005267834.1 | XP_005267891.1 | 6 | 3240 | ||
XM_017021426.1 | XP_016876915.1 | 8 | 3579 | ||
GenBank mRNA | AL137760.1 | - | 1 | 810 | |
AY358687.1 | - | 7 | 2884 |