Gene Transcripts

Gene Symbol
ZBTB4
Entrez Gene ID
57659
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
KAISO-L1, ZNF903
Chromosome Location
Chr. 17: 7459366 - 7484249 on Build GRCh38
UniGene ID
Hs.35096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128833.1 NP_001122305.1 4 3443
NM_020899.3 NP_065950.2 4 3519
XM_006721563.3 XP_006721626.1 4 3370
XM_006721564.2 XP_006721627.1 4 3350
XM_011523972.2 XP_011522274.1 4 3802
GenBank mRNA AB040971.2 - 4 3443
AK027143.1 - 1 593
AK122826.1 - 2 2771
AK122971.1 - 4 3343
AY302699.1 - 4 3359
BC033259.1 - 1 1065
BC043352.2 - 4 3468 6175382

View Full Product Details